Assay Details
| Gene Symbol: cca-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381028.1 | NP_001367692.1 | 26 - 27 | 5215 | 58 | |
| NM_001381029.1 | NP_001368577.1 | 26 - 27 | 5184 | 58 | ||
| NM_001381030.1 | NP_001367691.1 | 27 - 28 | 5338 | 58 | ||
| NM_001381031.2 | NP_001367690.1 | 26 - 27 | 5239 | 58 | ||
| NM_171733.5 | NP_741848.1 | 28 - 29 | 5280 | 58 | ||
| GenBank mRNA | AF368920.1 | - | 26 - 27 | 5143 | 58 | |
| AY313898.1 | - | 26 - 27 | 5194 | 58 | ||
| AY313899.1 | - | 27 - 28 | 5338 | 58 | ||
| AY313900.1 | - | 26 - 27 | 5239 | 58 | ||
| AY322480.1 | - | 26 - 27 | 5215 | 58 | ||
More Information
Set Membership: |
|

cca-1
Genomic Map