Assay Details
Gene Symbol: dim-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029249.6 | NP_001024420.1 | 10 - 11 | 1393 | 81 | |
NM_001029250.5 | NP_001024421.1 | 4 - 5 | 439 | 81 | ||
GenBank mRNA | AF500489.1 | - | 4 - 5 | 446 | 81 | |
AY095447.1 | - | 10 - 11 | 1412 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
