Assay Details
Gene Symbol: dim-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029249.6 | NP_001024420.1 | 11 - 12 | 1801 | 75 | |
NM_001029250.5 | NP_001024421.1 | 5 - 6 | 847 | 75 | ||
GenBank mRNA | AF500489.1 | - | 5 - 6 | 854 | 75 | |
AY095447.1 | - | 11 - 12 | 1820 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
