Assay Details
Gene Symbol: nhx-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029549.4 | NP_001024720.1 | 14 - 15 | 1702 | 75 | |
NM_001373485.2 | NP_001359642.1 | 14 - 15 | 1702 | 75 | ||
GenBank mRNA | AF497828.1 | - | 14 - 15 | 1723 | 75 | |
AF497829.1 | - | 14 - 15 | 1723 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
