Assay Details
| Gene Symbol: nhx-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373485.2 | NP_001359642.1 | 16 - 17 | 1976 | 77 | |
| GenBank mRNA | AF497829.1 | - | 16 - 17 | 1997 | 77 | |
More Information
Set Membership: |
|

nhx-5
Genomic Map