Assay Details
| Gene Symbol: nhx-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029549.4 | NP_001024720.1 | 17 - 18 | 2018 | 140 | |
| NM_001373485.2 | NP_001359642.1 | 18 - 19 | 2070 | 140 | ||
| GenBank mRNA | AF497828.1 | - | 17 - 18 | 2039 | 140 | |
| AF497829.1 | - | 18 - 19 | 2091 | 140 | ||
More Information
Set Membership: |
|

nhx-5
Genomic Map