Assay Details
| Gene Symbol: nhx-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029549.4 | NP_001024720.1 | 2 - 3 | 405 | 100 | |
| NM_001373485.2 | NP_001359642.1 | 2 - 3 | 405 | 100 | ||
| GenBank mRNA | AF497828.1 | - | 2 - 3 | 426 | 100 | |
| AF497829.1 | - | 2 - 3 | 426 | 100 | ||
More Information
Set Membership: |
|

nhx-5
Genomic Map