Assay Details
| Gene Symbol: sma-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375119.3 | NP_001361905.1 | 1 - 2 | 123 | 108 | |
| NM_001375124.1 | NP_001361885.1 | 1 - 2 | 123 | 108 | ||
| NM_001375125.2 | NP_001361884.1 | 1 - 2 | 113 | 108 | ||
| NM_001377619.1 | NP_001364627.1 | 1 - 2 | 117 | 108 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001375124.1, NM_001375125.2 transcripts.
Set Membership: |
|

sma-9
Genomic Map