Assay Details
| Gene Symbol: sma-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375119.3 | NP_001361905.1 | 21 | 5493 | 73 | |
| NM_001375120.2 | NP_001361887.1 | 20 - 21 | 6039 | 73 | ||
| NM_001375121.3 | NP_001361888.1 | 21 - 22 | 5940 | 73 | ||
| NM_001375124.1 | NP_001361885.1 | 21 - 22 | 6120 | 73 | ||
| NM_001375125.2 | NP_001361884.1 | 20 - 21 | 5536 | 73 | ||
| NM_001383639.1 | 21 - 22 | 5664 | 73 | |||
| NM_171771.6 | NP_741896.2 | 18 - 19 | 5511 | 73 | ||
| GenBank mRNA | AY390550.1 | - | 11 - 12 | 1830 | 73 | |
More Information
Set Membership: |
|

sma-9
Genomic Map