Assay Details
| Gene Symbol: sma-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375119.3 | NP_001361905.1 | 23 - 24 | 5982 | 62 | |
| NM_001375120.2 | NP_001361887.1 | 23 - 24 | 6528 | 62 | ||
| NM_001375121.3 | NP_001361888.1 | 24 - 25 | 6429 | 62 | ||
| NM_001375122.2 | NP_001361889.1 | 23 - 24 | 6270 | 62 | ||
| NM_001375124.1 | NP_001361885.1 | 24 - 25 | 6609 | 62 | ||
| NM_001375125.2 | NP_001361884.1 | 23 - 24 | 6025 | 62 | ||
| NM_171771.6 | NP_741896.2 | 21 - 22 | 6000 | 62 | ||
| GenBank mRNA | AY390550.1 | - | 14 - 15 | 2319 | 62 | |
More Information
Set Membership: |
|

sma-9
Genomic Map