Assay Details
Gene Symbol: scd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373531.3 | NP_001360646.1 | 2 - 3 | 463 | 74 | |
NM_001373534.4 | NP_001360782.1 | 3 | 531 | 74 | ||
NM_001373535.3 | NP_001360784.1 | 2 - 3 | 166 | 74 | ||
NM_001381120.2 | NP_001368763.1 | 1 - 2 | 65 | 74 | ||
NM_001381121.2 | NP_001368761.1 | 1 - 2 | 61 | 74 | ||
NM_001381122.2 | NP_001367017.1 | 1 - 2 | 61 | 74 | ||
NM_001381123.2 | NP_001368762.1 | 2 - 3 | 435 | 74 | ||
NM_171974.7 | NP_741912.1 | 3 - 4 | 514 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
