Assay Details
Gene Symbol: scd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373531.3 | NP_001360646.1 | 9 | 2783 | 77 | |
NM_001373534.4 | NP_001360782.1 | 12 | 2803 | 77 | ||
NM_001381121.2 | NP_001368761.1 | 7 - 8 | 2333 | 77 | ||
NM_001381122.2 | NP_001367017.1 | 7 - 8 | 2267 | 77 | ||
NM_001381123.2 | NP_001368762.1 | 8 - 9 | 2755 | 77 | ||
NM_171974.7 | NP_741912.1 | 9 - 10 | 2834 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
