Assay Details
Gene Symbol: scd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373531.3 | NP_001360646.1 | 11 | 3058 | 100 | |
NM_001373534.4 | NP_001360782.1 | 12 | 3078 | 100 | ||
NM_001373535.3 | NP_001360784.1 | 9 - 10 | 2653 | 100 | ||
NM_001381120.2 | NP_001368763.1 | 8 - 9 | 2504 | 100 | ||
NM_001381121.2 | NP_001368761.1 | 9 - 10 | 2569 | 100 | ||
NM_001381122.2 | NP_001367017.1 | 9 - 10 | 2542 | 100 | ||
NM_001381123.2 | NP_001368762.1 | 10 - 11 | 2991 | 100 | ||
NM_171974.7 | NP_741912.1 | 11 - 12 | 3109 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
