Assay Details
| Gene Symbol: madd-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001368549.3 | NP_001355453.1 | 7 - 8 | 1215 | 81 | |
| NM_001368550.3 | NP_001355370.1 | 7 - 8 | 1215 | 81 | ||
| NM_001375168.2 | NP_001362073.1 | 7 - 8 | 1212 | 81 | ||
| NM_001381143.1 | NP_001368709.1 | 5 - 6 | 2095 | 81 | ||
| NM_001383672.1 | 7 - 8 | 1205 | 81 | |||
| NM_171799.8 | NP_741928.1 | 5 - 6 | 3726 | 81 | ||
More Information
Set Membership: |
|

madd-3
Genomic Map