Assay Details
| Gene Symbol: stim-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171858.5 | NP_741074.1 | 8 - 9 | 1215 | 113 | |
| GenBank mRNA | DQ812088.1 | - | 8 - 9 | 1169 | 113 | |
More Information
Set Membership: |
|

stim-1
Genomic Map