Assay Details
| Gene Symbol: lon-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379712.1 | NP_001367873.1 | 2 - 3 | 322 | 110 | |
| NM_065765.9 | NP_498166.1 | 2 - 3 | 320 | 110 | ||
| NM_065766.7 | NP_498167.1 | 2 - 3 | 322 | 110 | ||
More Information
Set Membership: |
|

lon-1
Genomic Map