Assay Details
| Gene Symbol: akt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028474.7 | NP_001023645.1 | 2 - 3 | 264 | 83 | |
| NM_001028475.4 | NP_001023646.1 | 2 - 3 | 270 | 83 | ||
| NM_001028476.6 | NP_001023647.1 | 2 - 3 | 270 | 83 | ||
| GenBank mRNA | AF072379.1 | - | 2 - 3 | 264 | 83 | |
| AF072380.1 | - | 2 - 3 | 264 | 83 | ||
More Information
Set Membership: |
|

akt-1
Genomic Map