Assay Details
| Gene Symbol: akt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028474.7 | NP_001023645.1 | 8 - 9 | 1493 | 81 | |
| NM_001028475.4 | NP_001023646.1 | 8 - 9 | 1514 | 81 | ||
| NM_001028476.6 | NP_001023647.1 | 8 - 9 | 1395 | 81 | ||
| GenBank mRNA | AF072379.1 | - | 8 - 9 | 1493 | 81 | |
| AF072380.1 | - | 8 - 9 | 1508 | 81 | ||
More Information
Set Membership: |
|

akt-1
Genomic Map