Assay Details
| Gene Symbol: slo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029088.2 | NP_001024259.1 | 13 - 14 | 2247 | 119 | |
| NM_001269873.3 | NP_001256802.1 | 14 - 15 | 2268 | 119 | ||
| NM_001269875.4 | NP_001256804.1 | 14 - 15 | 2292 | 119 | ||
| NM_001269876.3 | NP_001256805.1 | 13 - 14 | 2208 | 119 | ||
| NM_001269878.3 | NP_001256807.1 | 14 - 15 | 2268 | 119 | ||
| NM_001269880.3 | NP_001256809.1 | 14 - 15 | 2247 | 119 | ||
| GenBank mRNA | AF431891.1 | - | 13 - 14 | 2208 | 119 | |
| JQ582463.1 | - | 14 - 15 | 2268 | 119 | ||
| JQ606809.1 | - | 14 - 15 | 2247 | 119 | ||
More Information
Set Membership: |
|

slo-1
Genomic Map