Assay Details
| Gene Symbol: ajm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330906.2 | NP_001317734.1 | 21 - 22 | 4320 | 80 | |
| NM_001330908.3 | NP_001317889.1 | 15 - 16 | 3500 | 80 | ||
| NM_001330909.3 | NP_001317890.1 | 15 - 16 | 3275 | 80 | ||
| NM_001330910.3 | NP_001317891.1 | 13 - 14 | 3143 | 80 | ||
| NM_001330911.3 | NP_001317888.1 | 12 - 13 | 3029 | 80 | ||
| NM_001330912.3 | NP_001317897.1 | 11 - 12 | 2713 | 80 | ||
| NM_001330913.3 | NP_001317898.1 | 10 - 11 | 2468 | 80 | ||
| NM_001330914.3 | NP_001317899.1 | 9 - 10 | 2396 | 80 | ||
| NM_001330915.3 | NP_001317736.1 | 7 - 8 | 2099 | 80 | ||
| NM_001381045.1 | NP_001368606.1 | 21 - 22 | 4283 | 80 | ||
| NM_001381046.2 | NP_001367383.1 | 20 - 21 | 4168 | 80 | ||
| NM_001381048.1 | NP_001367381.1 | 21 - 22 | 4320 | 80 | ||
More Information
Set Membership: |
|

ajm-1
Genomic Map