Assay Details
| Gene Symbol: ajm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330908.3 | NP_001317889.1 | 18 - 19 | 4186 | 68 | |
| NM_001330909.3 | NP_001317890.1 | 18 - 19 | 3961 | 68 | ||
| NM_001330910.3 | NP_001317891.1 | 16 - 17 | 3829 | 68 | ||
| NM_001330911.3 | NP_001317888.1 | 15 - 16 | 3715 | 68 | ||
| NM_001330912.3 | NP_001317897.1 | 14 - 15 | 3399 | 68 | ||
| NM_001330913.3 | NP_001317898.1 | 13 - 14 | 3154 | 68 | ||
| NM_001330914.3 | NP_001317899.1 | 12 - 13 | 3082 | 68 | ||
| NM_001330915.3 | NP_001317736.1 | 10 - 11 | 2785 | 68 | ||
| NM_001381045.1 | NP_001368606.1 | 24 - 25 | 4969 | 68 | ||
| NM_001381046.2 | NP_001367383.1 | 23 - 24 | 4854 | 68 | ||
| NM_001381048.1 | NP_001367381.1 | 24 - 25 | 5006 | 68 | ||
More Information
Set Membership: |
|

ajm-1
Genomic Map