Assay Details
| Gene Symbol: ajm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330906.2 | NP_001317734.1 | 10 - 11 | 1605 | 101 | |
| NM_001330908.3 | NP_001317889.1 | 4 - 5 | 785 | 101 | ||
| NM_001330909.3 | NP_001317890.1 | 4 - 5 | 560 | 101 | ||
| NM_001330910.3 | NP_001317891.1 | 2 - 3 | 428 | 101 | ||
| NM_001330911.3 | NP_001317888.1 | 1 - 2 | 314 | 101 | ||
| NM_001381045.1 | NP_001368606.1 | 10 - 11 | 1568 | 101 | ||
| NM_001381046.2 | NP_001367383.1 | 10 - 11 | 1576 | 101 | ||
| NM_001381047.2 | NP_001367382.1 | 10 - 11 | 1568 | 101 | ||
| NM_001381048.1 | NP_001367381.1 | 10 - 11 | 1605 | 101 | ||
More Information
Set Membership: |
|

ajm-1
Genomic Map