Assay Details
Gene Symbol: F49E2.5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029524.6 | NP_001024695.1 | 8 - 9 | 1861 | 73 | |
NM_001373430.3 | NP_001360758.1 | 8 - 9 | 1845 | 73 | ||
NM_001373432.4 | NP_001360760.1 | 8 - 9 | 1842 | 73 | ||
NM_001381054.2 | NP_001368747.1 | 8 - 9 | 1800 | 73 | ||
NM_001381058.1 | NP_001368748.1 | 6 - 7 | 1402 | 73 | ||
NM_001381059.1 | NP_001368749.1 | 6 - 7 | 1402 | 73 | ||
NM_001381060.1 | NP_001368750.1 | 6 - 7 | 1402 | 73 | ||
NM_001381061.2 | NP_001368751.1 | 6 - 7 | 1402 | 73 | ||
NM_001381069.1 | NP_001367019.1 | 6 - 7 | 1402 | 73 | ||
NM_001383625.1 | 8 - 9 | 1849 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
