Assay Details
Gene Symbol: nhx-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029549.4 | NP_001024720.1 | 16 - 17 | 1976 | 113 | |
GenBank mRNA | AF497828.1 | - | 16 - 17 | 1997 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
