Assay Details
Gene Symbol: ttyh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077502.7 | NP_509903.1 | 2 - 3 | 318 | 78 | |
NM_171973.9 | NP_741904.1 | 2 - 3 | 320 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
