Assay Details
Gene Symbol: ttyh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383646.1 | 4 - 5 | 819 | 95 | ||
NM_077502.7 | NP_509903.1 | 3 - 4 | 686 | 95 | ||
NM_171973.9 | NP_741904.1 | 3 - 4 | 688 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
