Assay Details
Gene Symbol: ttyh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383646.1 | 7 - 8 | 1342 | 121 | ||
NM_077502.7 | NP_509903.1 | 6 - 7 | 1209 | 121 | ||
NM_171973.9 | NP_741904.1 | 6 - 7 | 1211 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
