Assay Details
| Gene Symbol: scd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373531.3 | NP_001360646.1 | 6 | 2299 | 66 | |
| NM_001373534.4 | NP_001360782.1 | 7 | 2319 | 66 | ||
| NM_001373535.3 | NP_001360784.1 | 6 | 2002 | 66 | ||
| NM_001381120.2 | NP_001368763.1 | 4 - 5 | 1853 | 66 | ||
| NM_001381121.2 | NP_001368761.1 | 4 - 5 | 1849 | 66 | ||
| NM_001381123.2 | NP_001368762.1 | 5 - 6 | 2271 | 66 | ||
| NM_171974.7 | NP_741912.1 | 6 - 7 | 2350 | 66 | ||
More Information
Set Membership: |
|

scd-1
Genomic Map