Assay Details
Gene Symbol: gars-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382861.1 | 4 - 5 | 1195 | 65 | ||
NM_065692.6 | NP_498093.1 | 4 - 5 | 1244 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
