Assay Details
Gene Symbol: tlk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268088.3 | NP_001255017.1 | 4 - 5 | 2445 | 136 | |
NM_001382967.1 | 6 - 7 | 2806 | 136 | |||
NM_066744.5 | NP_499145.2 | 7 - 8 | 2876 | 136 | ||
NM_181913.5 | NP_871642.2 | 6 - 7 | 2688 | 136 | ||
GenBank mRNA | AY450852.1 | - | 6 - 7 | 2688 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
