Assay Details
Gene Symbol: mtm-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181951.6 | NP_871680.1 | 1 - 2 | 361 | 74 | |
GenBank mRNA | AY313179.1 | - | 1 - 2 | 344 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
