Assay Details
Gene Symbol: mtm-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027431.6 | NP_001022602.1 | 4 - 5 | 1266 | 101 | |
NM_067352.6 | NP_499753.1 | 4 - 5 | 1272 | 101 | ||
NM_181951.6 | NP_871680.1 | 4 - 5 | 1587 | 101 | ||
GenBank mRNA | AY313177.1 | - | 4 - 5 | 1252 | 101 | |
AY313179.1 | - | 4 - 5 | 1570 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
