Assay Details
Gene Symbol: mtm-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027431.6 | NP_001022602.1 | 7 - 8 | 1965 | 93 | |
NM_067352.6 | NP_499753.1 | 7 - 8 | 1971 | 93 | ||
NM_181951.6 | NP_871680.1 | 7 - 8 | 2286 | 93 | ||
GenBank mRNA | AY313177.1 | - | 7 - 8 | 1951 | 93 | |
AY313179.1 | - | 7 - 8 | 2269 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
