Assay Details
| Gene Symbol: hira-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065700.6 | NP_498101.2 | 7 - 8 | 1104 | 84 | |
| NR_151567.1 | 7 - 8 | 1104 | 84 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hira-1| Gene Symbol: hira-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065700.6 | NP_498101.2 | 7 - 8 | 1104 | 84 | |
| NR_151567.1 | 7 - 8 | 1104 | 84 | |||
Set Membership: |
|