Assay Details
| Gene Symbol: Y48G1C.10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306236.3 | NP_001293165.1 | 5 - 6 | 609 | 74 | |
| NM_182095.4 | NP_871895.2 | 5 - 6 | 605 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_182095.4 transcripts.
Set Membership: |
|

Y48G1C.10
Genomic Map