Assay Details
Gene Symbol: Y48G1C.10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306236.3 | NP_001293165.1 | 7 - 8 | 918 | 72 | |
NM_182095.4 | NP_871895.2 | 7 - 8 | 914 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_182095.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
