Assay Details
Gene Symbol: Y48G1C.10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306236.3 | NP_001293165.1 | 11 - 12 | 1456 | 81 | |
NM_182095.4 | NP_871895.2 | 11 - 12 | 1443 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
