Assay Details
| Gene Symbol: Y48G1C.10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306236.3 | NP_001293165.1 | 12 - 13 | 1677 | 155 | |
| NM_182095.4 | NP_871895.2 | 12 - 13 | 1664 | 155 | ||
More Information
Set Membership: |
|

Y48G1C.10
Genomic Map