Assay Details
| Gene Symbol: din-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267377.3 | NP_001254306.1 | 9 - 10 | 4115 | 99 | |
| NM_001381614.1 | NP_001367110.1 | 15 - 16 | 5271 | 99 | ||
| NM_064084.8 | NP_496485.2 | 14 - 15 | 5319 | 99 | ||
| NM_182101.5 | NP_871901.1 | 15 - 16 | 5249 | 99 | ||
More Information
Set Membership: |
|

din-1
Genomic Map