Assay Details
| Gene Symbol: din-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267377.3 | NP_001254306.1 | 10 - 11 | 4220 | 139 | |
| NM_001381614.1 | NP_001367110.1 | 16 - 17 | 5376 | 139 | ||
| NM_064083.6 | NP_496484.3 | 14 - 15 | 4834 | 139 | ||
| NM_064084.8 | NP_496485.2 | 15 - 16 | 5424 | 139 | ||
| NM_182101.5 | NP_871901.1 | 16 - 17 | 5354 | 139 | ||
More Information
Set Membership: |
|

din-1
Genomic Map