Assay Details
| Gene Symbol: din-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267377.3 | NP_001254306.1 | 13 - 14 | 5662 | 65 | |
| NM_001267378.3 | NP_001254307.1 | 1 - 2 | 178 | 65 | ||
| NM_001381614.1 | NP_001367110.1 | 19 - 20 | 6818 | 65 | ||
| NM_064083.6 | NP_496484.3 | 17 - 18 | 6276 | 65 | ||
| NM_064084.8 | NP_496485.2 | 18 - 19 | 6866 | 65 | ||
| NM_182101.5 | NP_871901.1 | 19 - 20 | 6796 | 65 | ||
| NM_182102.6 | NP_871902.1 | 2 - 3 | 2253 | 65 | ||
More Information
Set Membership: |
|

din-1
Genomic Map