Assay Details
| Gene Symbol: din-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267377.3 | NP_001254306.1 | 15 - 16 | 5946 | 66 | |
| NM_001267378.3 | NP_001254307.1 | 3 - 4 | 462 | 66 | ||
| NM_001381614.1 | NP_001367110.1 | 21 - 22 | 7102 | 66 | ||
| NM_064083.6 | NP_496484.3 | 19 - 20 | 6560 | 66 | ||
| NM_064084.8 | NP_496485.2 | 20 - 21 | 7150 | 66 | ||
| NM_182101.5 | NP_871901.1 | 21 - 22 | 7080 | 66 | ||
| NM_182102.6 | NP_871902.1 | 4 - 5 | 2537 | 66 | ||
More Information
Set Membership: |
|

din-1
Genomic Map