Assay Details
Gene Symbol: din-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381614.1 | NP_001367110.1 | 4 - 5 | 448 | 64 | |
NM_064083.6 | NP_496484.3 | 3 - 4 | 494 | 64 | ||
NM_064084.8 | NP_496485.2 | 3 - 4 | 496 | 64 | ||
NM_182101.5 | NP_871901.1 | 4 - 5 | 426 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381614.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
