Assay Details
| Gene Symbol: din-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267377.3 | NP_001254306.1 | 2 - 3 | 294 | 96 | |
| NM_001381614.1 | NP_001367110.1 | 8 - 9 | 1450 | 96 | ||
| NM_064083.6 | NP_496484.3 | 7 - 8 | 1496 | 96 | ||
| NM_064084.8 | NP_496485.2 | 7 - 8 | 1498 | 96 | ||
| NM_182101.5 | NP_871901.1 | 8 - 9 | 1428 | 96 | ||
More Information
Set Membership: |
|

din-1
Genomic Map