Assay Details
| Gene Symbol: F10G7.10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381399.2 | NP_001366635.1 | 10 - 11 | 1634 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F10G7.10| Gene Symbol: F10G7.10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381399.2 | NP_001366635.1 | 10 - 11 | 1634 | 70 | |
Set Membership: |
|