Assay Details
| Gene Symbol: cpna-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062335.7 | NP_494736.1 | 2 - 3 | 422 | 68 | |
| NM_062336.5 | NP_494737.1 | 2 - 3 | 422 | 68 | ||
| NM_182185.6 | NP_871985.1 | 2 - 3 | 424 | 68 | ||
More Information
Set Membership: |
|

cpna-1
Genomic Map