Assay Details
| Gene Symbol: cpna-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182185.6 | NP_871985.1 | 5 - 6 | 1361 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cpna-1| Gene Symbol: cpna-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182185.6 | NP_871985.1 | 5 - 6 | 1361 | 66 | |
Set Membership: |
|