Assay Details
| Gene Symbol: ctns-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063303.6 | NP_495704.1 | 1 - 2 | 145 | 136 | |
| NM_182222.6 | NP_872022.1 | 1 - 2 | 147 | 136 | ||
More Information
Set Membership: | 
		   
					
						
							 | 
				

ctns-1
Genomic Map