Assay Details
| Gene Symbol: ctns-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063303.6 | NP_495704.1 | 4 - 5 | 866 | 108 | |
| NM_182222.6 | NP_872022.1 | 4 - 5 | 868 | 108 | ||
More Information
Set Membership: |
|

ctns-1
Genomic Map