Assay Details
Gene Symbol: sra-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182233.2 | NP_872033.2 | 3 - 4 | 364 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
